Learn More
Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Drugs of Abuse Assay (CJ&F)
Shop All Thermo Scientific ProductsDescription
Determine the presence of mitragynine, the major alkaloid found in leaf extracts of the Kratom plant Mitragyna speciosa, in human urine using this automated immunoassay. CJ&F only.
Specifically detect mitragynine, the major alkaloid found in leaf extracts of the Kratom plant Mitragyna speciosa, in human urine using this automated immunoassay. For Criminal Justice and Forensic use only in the USA.
- Specifically detects mitragynine, the major alkaloid found in leaf extracts of the Kratom plant, Mitragyna speciosa
- High correlation to LC-MS/MS
- Qualitative and semi-quantitative detection
- Applications available on a wide range of clinical analyzers
- Available April 2019
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | CEDIA Mitragynine Control Set |
| Detectable Analytes | Mitragynine |
| DoA Calibrators | Mitragynine Calibrator, Negative Calibrator II |
| Sample Type | Urine |
| Type | Mitragynine (Kratom) Assay |
| Quantity | 1 x 65 mL |
For sale in the U.S.A. only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.