Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F)

Catalog No. 22000199
Change view
Click to view available options
Quantity:
1 x 5 mL
2 x 5 mL
Type:
Calibrator
Control
Concentration:
100 ng/mL
20 ng/mL
200 ng/mL
50 ng/mL
Low 37.5 ng/mL, High 62.5 ng/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Concentration
22-000-199 1 x 5 mL Calibrator 20 ng/mL
22-000-200 1 x 5 mL Calibrator 50 ng/mL
22-000-201 1 x 5 mL Calibrator 100 ng/mL
22-000-202 1 x 5 mL Calibrator 200 ng/mL
22-000-203 2 x 5 mL Control Low 37.5 ng/mL, High 62.5 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.
Catalog No. 22-000-199 Supplier Thermo Scientific™ Supplier No. 10026590
Add to Cart
Add to Cart
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.

Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay using the Thermo Scientific CEDIA Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F only).

Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay and ensure accuracy of results using the Thermo Scientific CEDIA Mitragynine (Kratom) Calibrators and Controls (CJ&F). The five calibrators (including negative) establish a standard curve that correlates the instrument signals with the calibrators' respective concentrations in ng/mL. This allows for semi-quantitative measurements. Two controls, one above and one below the cutoff (50ng/mL), validate the calibration curve. The cutoff calibrator provides a reference point for qualitative detection of positive or negative results. For Criminal Justice and Forensic use only in the USA.

  • Ensures assay accuracy
  • Supports qualitative and semi-quantitative measurements
  • Available April 2019
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Concentration 20 ng/mL
Detectable Analytes Mitragynine
Sample Type Urine
Type Calibrator
Quantity 1 x 5 mL

For sale in the U.S.A. only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.