Learn More
Thermo Scientific™ CEDIA Mitragynine (Kratom) 50ng/mL Calibrator
Catalog No. 09980604 Shop All Thermo Scientific Products
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.
Catalog No. 09-980-604 Supplier Thermo Scientific™ Supplier No. 10026591/DEL
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.
Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay using the Thermo Scientific CEDIA Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F only).
Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay and ensure accuracy of results using the Thermo Scientific CEDIA Mitragynine (Kratom) Calibrators and Controls (CJ&F). The five calibrators (including negative) establish a standard curve that correlates the instrument signals with the calibrators' respective concentrations in ng/mL. This allows for semi-quantitative measurements. Two controls, one above and one below the cutoff (50ng/mL), validate the calibration curve. The cutoff calibrator provides a reference point for qualitative detection of positive or negative results. For Criminal Justice and Forensic use only in the USA.
  • Ensures assay accuracy
  • Supports qualitative and semi-quantitative measurements
  • Available April 2019
TRUSTED_SUSTAINABILITY
For Criminal Justice and Forensic Use Only.