Learn More
Thermo Scientific™ CEDIA Mitragynine (Kratom) 100ng/mL Calibrator
Catalog No. 09980605 Shop All Thermo Scientific Products
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.
Catalog No. 09-980-605 Supplier Thermo Scientific™ Supplier No. 10026592/DEL
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.
Validate the CEDIA Mitragynine (Kratom) Assay using the CEDIA Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F only).
Validate the CEDIA Mitragynine (Kratom) Assay and ensure accuracy of results using the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Calibrators and Controls (CJ&F). The five calibrators (including negative) establish a standard curve that correlates the instrument signals with the calibrators' respective concentrations in ng/mL. This allows for semi-quantitative measurements. Two controls, one above and one below the cutoff (50ng/mL), validate the calibration curve. The cutoff calibrator provides a reference point for qualitative detection of positive or negative results. For Criminal Justice and Forensic use only in the USA.
  • Ensures assay accuracy
  • Supports qualitative and semi-quantitative measurements
  • Available April 2019
TRUSTED_SUSTAINABILITY
For Criminal Justice and Forensic Use Only.