Learn More
Fine Science Tools Dissector Scissors - Heavy Blades
SDP
Catalog No. NC9872127
Encompass
Tip Shape: Angled to Side/ Tips: Sharp-Sharp/ Cutting Edge: 12mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No. Heavy blades for cutting fine bone.
Catalog No. NC9872127 Supplier Fine Science Tools Supplier No. 1408209
Tip Shape: Angled to Side/ Tips: Sharp-Sharp/ Cutting Edge: 12mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No. Heavy blades for cutting fine bone.