Learn More
Thermo Scientific STRONG SOLVENT LOOP
Catalog No. NC1836544 Shop All Thermo Scientific Products
Encompass
Solvent resistant tubing forms continuous recirculation pathway for aggressive solvents in closed system without air exposure for use with pumps and equipment.
Catalog No. NC1836544 Supplier Thermo Scientific Supplier No. 6036.2200
Solvent resistant tubing forms continuous recirculation pathway for aggressive solvents in closed system without air exposure for use with pumps and equipment.