Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium bromide, ultra dry, 99.995% (metals basis)

Catalog No. AA3571706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3571706 5 g
AA3571703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3571706 Supplier Thermo Scientific Chemicals Supplier No. 03571706
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10476-81-0 | Br2Sr | 247.428 g/mol

It is used as sedatives in medicine and analytical reagents, for facile synthesis of poly(phenylcarbyne), which is a precursor to diamond-like carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-81-0
Molecular Formula Br2Sr
Molecular Weight (g/mol) 247.428
MDL Number MFCD00049548
InChI Key YJPVTCSBVRMESK-UHFFFAOYSA-L
Synonym acmc-20ajmt, strontium bromide srbr2, strontium 2+ ion dibromide
PubChem CID 5360351
IUPAC Name strontium;dibromide
SMILES [Br-].[Br-].[Sr+2]

Specifications

Melting Point 643°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula SrBr2
Merck Index 14,8837
Solubility Information Soluble in water,carbinol,ethanol,amyl alcohol. Insoluble in ether.
Formula Weight 247.44
Percent Purity 99.995%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.216 g/mL
Assay (metals basis)
Chemical Name or Material Strontium bromide, Ultra dry
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-969-5
RTECSNumber WK8050000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.