Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Strontium carbonate

Catalog No. 289833250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-983-3250G 250 g
28-983-31KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-983-3250G Supplier Sigma Aldrich Supplier No. 289833250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Strontium carbonate is insoluble in water. It is used predominantly in producing other strontium salts.Strontium carbonate is used to prepare: Barium strontium titanate Barium strontium titanate thin films It may be used to prepare europium doped di-strontium magnesium di-silicate phosphor. Strontium carbonate is used in pyrotechnics, ceramic ferrites, inexpensive fireworks colorant.

Specifications

CAS 1633-05-2
Quantity 250 g
Molecular Formula CO3Sr
Linear Formula SrCO3
MDL Number MFCD00011250
Molecular Weight (g/mol) 147.63
Percent Purity ≥98%
Density 3.7 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.