Learn More
Strontium carbonate, 97.5%
Description
As a catalyst, in radiation-resistant glass, in ceramic ferrites, and in pyrotechnicsUsed as a colorant in fireworks. It is also used in ceramics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1100°C (decomposition) |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | 97.5% |
| Solubility Information | Practically insoluble in water. Soluble in acids and carbonated waterSoluble in ammonium chloride. Slightly soluble in ammonia and water. |
| Formula Weight | 147.63 |
| Density | 3.70 g/mL |
| Chemical Name or Material | Strontium carbonate |
Safety and Handling
| EINECSNumber | 216-643-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.