Learn More
Strem, An Ascensus Company CAS# 1633-05-2. 250g. Strontium carbonate, min. 97% (contains ~1.0-2.5% barium carbonate). MFCD00011250
SDP
Catalog No. 5090115172
Encompass
CAS# 1633-05-2. 250g. Strontium carbonate, min. 97% (contains ~1.0-2.5% barium carbonate). MFCD00011250. Molecular Weight: 147.63. Molecular Formula: SrCO3. Color/form: white pwdr. Strem# 93-3804. http://www.strem.com/catalog/v/93-3804/
Catalog No. 50-901-15172 Supplier Strem, An Ascensus Company Supplier No. 933804250G
CAS# 1633-05-2. 250g. Strontium carbonate, min. 97% (contains ~1.0-2.5% barium carbonate). MFCD00011250. Molecular Weight: 147.63. Molecular Formula: SrCO3. Color/form: white pwdr. Strem# 93-3804. http://www.strem.com/catalog/v/93-3804/