Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium carbonate, Puratronic™, 99.994% (metals basis)

Catalog No. AA3661814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3661814 25 g
AA3661822 100 g
AA3661836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3661814 Supplier Thermo Scientific Chemicals Supplier No. 03661814
Add to Cart

Used as a colorant in fireworks. It is also used in ceramics industry. | CAS: 1633-05-2 | CO3Sr | 147.63 g/mol

Used as a colorant in fireworks. It is also used in ceramics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-05-2
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
Synonym strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
PubChem CID 15407
IUPAC Name strontium;carbonate
SMILES [Sr++].[O-]C([O-])=O

Specifications

Melting Point 1100°C (decomposition)
Physical Form Powder
Quantity 25 g
Solubility Information Soluble in ammonium chloride. Slightly soluble in ammonia and water.
Formula Weight 147.63
Percent Purity 99.994%
Packaging Plastic bottle
Specific Gravity 3.7
Density 3.7
Chemical Name or Material Strontium carbonate
EINECSNumber 216-643-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.