Learn More
Medchemexpress LLC Strontium chloride, anhydrous grade, metals basis | 10476-85-4 | 99.99% | 158.53 | 50 MG
SDP
Catalog No. 5000401131
Encompass_Preferred
Strontium chloride, anhydrous grade, is a high-purity biochemical reagent with 99.99% metals basis. It is provided as a solid and is soluble in H2O at 100 mg/mL. This product is intended for research purposes only and not for human or patient use.
  • Provided as a solid appearance
  • Soluble in H2O
  • Suitable for research applications
  • Controlled substance with potential sales restrictions in certain territories
Catalog No. 50-004-01131 Supplier Medchemexpress LLC Supplier No. HY22009A50MG
Strontium chloride, anhydrous grade, is a high-purity biochemical reagent with 99.99% metals basis. It is provided as a solid and is soluble in H2O at 100 mg/mL. This product is intended for research purposes only and not for human or patient use.
  • Provided as a solid appearance
  • Soluble in H2O
  • Suitable for research applications
  • Controlled substance with potential sales restrictions in certain territories