Learn More
Medchemexpress LLC Strontium chloride, anhydrous grade, metals basis | 10476-85-4 | MFCD00011249 | 99.99% | 158.53 | 100 MG
SDP
Catalog No. 5000401147
Encompass_Preferred
Strontium chloride, anhydrous grade, 99.99% metals basis is a biochemical reagent suitable for various laboratory applications.
  • Used as a biochemical reagent
  • Suitable for laboratory chemical applications
  • Can be used for the manufacture of substances
  • Soluble in water at 100 mg/mL
Catalog No. 50-004-01147 Supplier Medchemexpress LLC Supplier No. HY22009A100MG
Strontium chloride, anhydrous grade, 99.99% metals basis is a biochemical reagent suitable for various laboratory applications.
  • Used as a biochemical reagent
  • Suitable for laboratory chemical applications
  • Can be used for the manufacture of substances
  • Soluble in water at 100 mg/mL