Learn More
Strem, An Ascensus Company CAS# 1311-10-0. 500g. Strontium hydroxide octahydrate, tech. gr.. MFCD00150395
SDP
Catalog No. 5090115180
Encompass
CAS# 1311-10-0. 500g. Strontium hydroxide octahydrate, tech. gr.. MFCD00150395. Molecular Weight: 121.63 (265.76). Molecular Formula: Sr(OH)2·8H2O. Color/form: white xtl. Strem# 93-3810. http://www.strem.com/catalog/v/93-3810/
Catalog No. 50-901-15180 Supplier Strem, An Ascensus Company Supplier No. 933810500G
CAS# 1311-10-0. 500g. Strontium hydroxide octahydrate, tech. gr.. MFCD00150395. Molecular Weight: 121.63 (265.76). Molecular Formula: Sr(OH)2·8H2O. Color/form: white xtl. Strem# 93-3810. http://www.strem.com/catalog/v/93-3810/