Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Strontium hydroxide octahydrate

Catalog No. 463752100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-375-2100G 100 g
46-375-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-375-2100G Supplier Sigma Aldrich Supplier No. 463752100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Strontium hydroxide octahydrate

Specifications

CAS 1311-10-0
Quantity 100 g
Molecular Formula H2O2Sr -+ 8H2O
Linear Formula Sr(OH)2 -+ 8H2O
MDL Number MFCD00150395
Synonym Strontium dihydroxide octahydrate; Strontium hydrate
Molecular Weight (g/mol) 265.76
Percent Purity 99.995% trace metals basis
Density 1.9 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.