Learn More
Strontium hydroxide octahydrate, tech.
Description
Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C-8H2O |
| Physical Form | Crystalline |
| Quantity | 50 g |
| Solubility Information | Soluble in hot water and acid. Insoluble in acetone. |
| Formula Weight | 265.76 (121.60 Anhydrous) |
| Grade | Technical |
| Sensitivity | Air sensitive; Hygroscopic |
| Density | 1.9 |
| Chemical Name or Material | Strontium hydroxide octahydrate |
Sécurité et manipulation
| Hazard Category | H314-H335 |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 242-367-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.