Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium molybdenum oxide, 99%

Catalog No. AA4023809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4023809 10 g
AA4023818 50 g
AA4023830 250 g
AA40238A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4023809 Supplier Thermo Scientific Chemicals Supplier No. 04023809
Add to Cart

CAS: 13470-04-7 | SrMoO4

It is used as an optical material for a small range of special applications, for example, as an optical coating on lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13470-04-7
Molecular Formula SrMoO4
MDL Number MFCD00049550
Synonym Strontium molybdate

Specifications

Melting Point (decomposition)
Color Colorless
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (excluding Ba)
Solubility Information Insoluble in water. Soluble in acid
Formula Weight 247.56
Density 4.54
Chemical Name or Material Strontium molybdenum oxide, Ba < 0.3%
EINECSNumber 236-730-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.