Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium nitrate, Puriss. p.a., ACS Reagent, ≥99.0% (complexometric), Solstice

Catalog No. 6001562 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-015-62 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-015-62 Supplier Solstice Supplier No. 31633250G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent,≥99.0% (complexometric)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
IUPAC Name strontium;dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]

Specifications

Quantity 250 g
Linear Formula Sr(NO3)2
UN Number 1507
Formula Weight 211.63g/mol
Percent Purity ≥99% (complexometric)
Grade ACS/puriss. p.a.
Chemical Name or Material Strontium Nitrate
Hazard Statement P210-P220-P221-P280-P305 + P351 + P338 + P310-P370 + P378
Precautionary Statement H272-H302-H318
EINECSNumber 233-131-9
RTECSNumber WK9800000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.