Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium perchlorate trihydrate, 98%

Catalog No. p-4407971
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4557709 10 g
AA4557722 100 g
AA4557736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4557709 Supplier Thermo Scientific Chemicals Supplier No. 04557709
Only null left
Add to Cart

CAS: 15650-09-6 | Cl2H6O11Sr | 340.56 g/mol

Strontium perchlorate trihydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15650-09-6
Molecular Formula Cl2H6O11Sr
Molecular Weight (g/mol) 340.56
MDL Number MFCD09039117
InChI Key VRRVWIQWZWFYDJ-UHFFFAOYSA-L
Synonym strontium perchlorate trihydrate, 2clo4.sr.3h2o, strontium perchlorate, trihydrate, strontium 2+ trihydrate diperchlorate
PubChem CID 71435311
SMILES O.O.O.[Sr++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 10 g
Assay Percent Range 98%
Solubility Information Very soluble in water. Soluble in alcohol.
IUPAC Name strontium;diperchlorate;trihydrate
Formula Weight 340.54 (286.52 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Strontium perchlorate trihydrate
Hazard Category H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.