Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Strontium peroxide, 12.3% available oxygen

Manufacturer:  Alfa Aesar™ 4631030

Quantity 250g

 View more versions of this product

Catalog No. AA4631030


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1314-18-7
SrO2
MFCD00084686
14,8847
O2Sr
119.62
UN1509
Strontium peroxide, 12.3% available oxygen

Specifications

1314-18-7
O2Sr
119.62
UN1509
−100 Mesh
Pale brown
250g
Powder
SrO2
MFCD00084686
14,8847
Moisture Sensitive
215°C (dec.)
Strontium peroxide, 12.3% available oxygen

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H318-H335-H315
May intensify fire
oxidizer.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: STRONTIUM PEROXIDE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit