Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium Plasma Grade Reference Standard, 1mL = 1mg Sr, 1000ppm Sr, Ricca Chemical
SDP

Catalog No. 81014 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81014 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 81014 Supplier Ricca Chemical Company Supplier No. 81014

Chemical Identifiers

CAS 10042-76-9 , 7697-37-2 , 7732-18-5
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly natural
Concentration or Composition Notes 1000ppm Sr (Sr(NO3)2 in 5% HNO3)
Equivalence 1 mL = 1 mg Sr
Physical Form Liquid
Color Colorless
Quantity 120 mL
Chemical Name or Material Strontium Plasma Grade Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.