Learn More
Medchemexpress LLC HY-17397 100mg Medchemexpress, Strontium Ranelate CAS:135459-87-9 Purity:>98%
SDP
Catalog No. 502081942
Encompass_Preferred
Medchemexpress, HY-17397 100mg Strontium Ranelate CAS:135459-87-9 Strontium Ranelate (S12911) is an antiosteoporotic agent that acts by reducing bone resorption and promoting bone formation, thereby inducing a positive bone balance. Strontium Ranelate also can activate the calcium-sensing receptor (CaSR) in non skeletal cells, resulting in the activation of inositol 1, 4, 5-triphosphate production and mitogen-activated protein kinase signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1942 Supplier Medchemexpress LLC Supplier No. HY17397100MG
Medchemexpress, HY-17397 100mg Strontium Ranelate CAS:135459-87-9 Strontium Ranelate (S12911) is an antiosteoporotic agent that acts by reducing bone resorption and promoting bone formation, thereby inducing a positive bone balance. Strontium Ranelate also can activate the calcium-sensing receptor (CaSR) in non skeletal cells, resulting in the activation of inositol 1, 4, 5-triphosphate production and mitogen-activated protein kinase signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.