Learn More
Medchemexpress LLC Strontium Ranelate | 135459-87-9 | 99.99% | 513.49 | 5 G
SDP
Catalog No. 5000385642
Encompass_Preferred
Strontium Ranelate is an antiosteoporotic agent that reduces bone resorption and promotes bone formation, leading to a positive bone balance. It activates the calcium-sensing receptor (CaSR) in non-skeletal cells, initiating inositol 1, 4, 5-triphosphate production and mitogen-activated protein kinase signaling. For research use only.
  • Reduces bone resorption and promotes bone formation.
  • Induces a positive bone balance.
  • Activates calcium-sensing receptor (CaSR) in non-skeletal cells.
  • Increases early and late osteoblast markers (ALP, OCN) in mouse calvaria cells in vitro.
  • Increases alkaline phosphatase activity and prostaglandin E2 production.
  • Increases bone mass in vertebrae and lumbar/femur in adult mice and rats in vivo.
  • Decreases bone resorption and increases bone formation in alveolar bone.
  • Prevents trabecular bone loss in ovariectomized rats, improving bone strength.
Catalog No. 50-003-85642 Supplier Medchemexpress LLC Supplier No. HY173975G
Strontium Ranelate is an antiosteoporotic agent that reduces bone resorption and promotes bone formation, leading to a positive bone balance. It activates the calcium-sensing receptor (CaSR) in non-skeletal cells, initiating inositol 1, 4, 5-triphosphate production and mitogen-activated protein kinase signaling. For research use only.
  • Reduces bone resorption and promotes bone formation.
  • Induces a positive bone balance.
  • Activates calcium-sensing receptor (CaSR) in non-skeletal cells.
  • Increases early and late osteoblast markers (ALP, OCN) in mouse calvaria cells in vitro.
  • Increases alkaline phosphatase activity and prostaglandin E2 production.
  • Increases bone mass in vertebrae and lumbar/femur in adult mice and rats in vivo.
  • Decreases bone resorption and increases bone formation in alveolar bone.
  • Prevents trabecular bone loss in ovariectomized rats, improving bone strength.