Learn More
Selleck Chemical LLC Strontium Ranelate S2050-50mg
SDP
Catalog No. 503111268
Encompass_Preferred
Strontium Ranelate is a strontium(II) salt of ranelic acid for (-)-desmethoxyverapamil binding to calcium channel with IC50 of 0 5 mM
Catalog No. 50-311-1268 Supplier Selleck Chemical LLC Supplier No. S205050MG
Strontium Ranelate is a strontium(II) salt of ranelic acid for (-)-desmethoxyverapamil binding to calcium channel with IC50 of 0 5 mM