Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium Ranelate Hydrate 97.0+%, TCI America™
SDP

Catalog No. S09381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09381G Supplier TCI America Supplier No. S09381G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 674773-07-0
Molecular Formula C12H8N2O9SSr2
Molecular Weight (g/mol) 531.501
MDL Number MFCD09038742
InChI Key KXBBXBBUNMMNSU-UHFFFAOYSA-J
Synonym Ranelic Acid Strontium Salt
PubChem CID 134128914
IUPAC Name distrontium;5-[bis(carboxylatomethyl)amino]-3-(carboxylatomethyl)-4-cyanothiophene-2-carboxylate;hydrate
SMILES C(C1=C(SC(=C1C#N)N(CC(=O)[O-])CC(=O)[O-])C(=O)[O-])C(=O)[O-].O.[Sr+2].[Sr+2]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 513.49
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Strontium Ranelate Hydrate
RTECSNumber XM7581000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.