Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium Standard, 1mL = 1mg Sr, 1000ppm Sr, Ricca Chemical
SDP

Catalog No. 81004 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81004 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 81004 Supplier Ricca Chemical Company Supplier No. 81004

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1633-05-2 , 7697-37-2 , 7732-18-5
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
PubChem CID 15407
IUPAC Name strontium(2+) carbonate
SMILES [Sr++].[O-]C([O-])=O

Specifications

Packaging Poly natural
Concentration or Composition Notes 1,000ppm Sr
Equivalence 1 mL = 1 mg Sr
Physical Form Liquid
Color Colorless
Quantity 120 mL
Chemical Name or Material Strontium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.