Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium titanium oxide, 99+% (metals basis)

Catalog No. p-7048751
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1139930 250 g
AA11399A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1139930 Supplier Thermo Scientific Chemicals Supplier No. 01139930
Only null left
Add to Cart

Used in electronics and in electrical insulation | CAS: 12060-59-2 | O3SrTi | 183.48 g/mol

Strontium titanium oxide is used as a diamond stimulant and also used as a strontium-90 in radioisotope thermoelectric generators. It is used in the electronics and electrical insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
IUPAC Name strontium;dioxido(oxo)titanium
SMILES [Sr++].[O-][Ti]([O-])=O

Specifications

Melting Point 2060°C
Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water and most solvents.
Formula Weight 183.51
Percent Purity ≥99%
Odor Odorless
Density 4.810 g/mL
Assay (metals basis)
Chemical Name or Material Strontium titanium oxide
EINECSNumber 235-044-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.