Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Strontium titanium oxide, 99+% (metals basis)

Used in electronics and in electrical insulation

Manufacturer:  Alfa Aesar™ 1139930

Quantity 250g

 View more versions of this product

Catalog No. AA1139930


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12060-59-2
MFCD00049554
Strontium titanium oxide
183.51
Strontium titanate

Specifications

12060-59-2
183.51
Strontium titanate
4.810g/mL
2060°C
Strontium titanium oxide
Powder
MFCD00049554
Insoluble in water and most solvents
Odorless
250g
≥99%, (metals basis)

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit