Learn More
Medchemexpress LLC Strophanthidin 25mg | 66-28-4 | 25 MG
SDP
Catalog No. 5000228913
Encompass_Preferred
Strophanthidin is a naturally occurring cardiac glycoside used as a research standard and Na+/K+-ATPase modulator. It is supplied as a solid suitable for analytical, in vitro, and in vivo studies, with recommended solvents and storage guidance to maintain stability.
  • Naturally occurring cardiac glycoside and Na+/K+-ATPase modulator.
  • High purity suitable for analytical and biological assays.
  • Offered in small research quantities for assay development.
  • Soluble in ethanol; in vivo formulation protocols are provided for clear dosing solutions.
  • Store solid at 4°C and protect from moisture and light to preserve integrity.
Catalog No. 50-002-28913 Supplier Medchemexpress LLC Supplier No. HY11425225MG
Strophanthidin is a naturally occurring cardiac glycoside used as a research standard and Na+/K+-ATPase modulator. It is supplied as a solid suitable for analytical, in vitro, and in vivo studies, with recommended solvents and storage guidance to maintain stability.
  • Naturally occurring cardiac glycoside and Na+/K+-ATPase modulator.
  • High purity suitable for analytical and biological assays.
  • Offered in small research quantities for assay development.
  • Soluble in ethanol; in vivo formulation protocols are provided for clear dosing solutions.
  • Store solid at 4°C and protect from moisture and light to preserve integrity.