Learn More
Medchemexpress LLC Strophanthidin | 5MG
SDP
Catalog No. 5000229049
Encompass_Preferred
Strophanthidin is a naturally available cardiac glycoside[1] Strophanthidin 0 1 and 1 nmol/L increases and 1 100 mol/L inhibits the Na /K -ATPase activities but Strophanthidin 10 and 100 nmol/L does not affect Na /K -ATPase activities in cardiac sarcolemmal[2] Strophanthidin increases both diastolic and systolic intracellular Ca2 concentration[3]
Catalog No. 50-002-29049 Supplier Medchemexpress LLC Supplier No. HY1142525MG
Strophanthidin is a naturally available cardiac glycoside[1] Strophanthidin 0 1 and 1 nmol/L increases and 1 100 mol/L inhibits the Na /K -ATPase activities but Strophanthidin 10 and 100 nmol/L does not affect Na /K -ATPase activities in cardiac sarcolemmal[2] Strophanthidin increases both diastolic and systolic intracellular Ca2 concentration[3]