Learn More
Cayman Chemical Strophanthidin, Purity: ≥90%, CAS: 66-28-4, 100 mg
SDP
Catalog No. 502917176
Encompass_Preferred
A cardenolide with diverse biological activities; inhibits N-acetyltransferase (IC50 = ~1.2 µM); reduces maximal Na+/K+-ATPase pump activity and cell attachment and increases cell death in MDCK-C7 cells at 1 µM; decreases the levels of SRC-3 in MCF-7 cells at 500 nM; increases calcium efflux and sarcoplasmic reticulum calcium levels in a voltage-clamp assay using isolated and perfused guinea pig hearts at 50 µM
Catalog No. 50-291-7176 Supplier Cayman Chemical Supplier No. 39244100 MG
A cardenolide with diverse biological activities; inhibits N-acetyltransferase (IC50 = ~1.2 µM); reduces maximal Na+/K+-ATPase pump activity and cell attachment and increases cell death in MDCK-C7 cells at 1 µM; decreases the levels of SRC-3 in MCF-7 cells at 500 nM; increases calcium efflux and sarcoplasmic reticulum calcium levels in a voltage-clamp assay using isolated and perfused guinea pig hearts at 50 µM