Learn More
Cayman Chemical StrophnthIdIn 50mg
SDP
Catalog No. 502916882
Encompass_Preferred
A cardenolide with diverse biological activities; inhibits N-acetyltransferase (IC50 = ~1.2 µM); reduces maximal Na+/K+-ATPase pump activity and cell attachment and increases cell death in MDCK-C7 cells at 1 µM; decreases the levels of SRC-3 in MCF-7 cells at 500 nM; increases calcium efflux and sarcoplasmic reticulum calcium levels in a voltage-clamp assay using isolated and perfused guinea pig hearts at 50 µM
Catalog No. 50-291-6882 Supplier Cayman Chemical Supplier No. 3924450 MG
A cardenolide with diverse biological activities; inhibits N-acetyltransferase (IC50 = ~1.2 µM); reduces maximal Na+/K+-ATPase pump activity and cell attachment and increases cell death in MDCK-C7 cells at 1 µM; decreases the levels of SRC-3 in MCF-7 cells at 500 nM; increases calcium efflux and sarcoplasmic reticulum calcium levels in a voltage-clamp assay using isolated and perfused guinea pig hearts at 50 µM