Learn More
Chemglass Life Sciences Stirrer, Magnetic, Five Position, Dura-Mag, 120V, with Factory Installed Alarm
SDP
Catalog No. 50143347
Encompass
Stirrer, Magnetic, Five Position, Dura-Mag, 120V, with Factory Installed Alarm. Dura-Mag™ digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment. Dura-Mag™ stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Optional factory installed motion alarm available. Speed range: 25-400RPM.
Catalog No. 50-143-347 Supplier Chemglass Life Sciences Supplier No. CLS4100A6
Stirrer, Magnetic, Five Position, Dura-Mag, 120V, with Factory Installed Alarm. Dura-Mag™ digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment. Dura-Mag™ stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Optional factory installed motion alarm available. Speed range: 25-400RPM.