Learn More
MOUSER ELECTRONICS Circular Push Pull Connectors Straight Plug C27 Collet 50 Ohm Solder
Catalog No. NC1887483
Encompass
00 Series Circular Push Pull Connectors were conceived for all applications in which a high density of connectors is necessary, especially for patch panels. The LEMO special self-latching system makes it possible to connect and disconnect with a simple axial push-pull, thereby reducing the space needed to mount sockets to an absolute minimum, up to 50 sockets per square decimeter.
Catalog No. NC1887483 Supplier MOUSER ELECTRONICS Supplier No. 736FFA00250CTAC27
00 Series Circular Push Pull Connectors were conceived for all applications in which a high density of connectors is necessary, especially for patch panels. The LEMO special self-latching system makes it possible to connect and disconnect with a simple axial push-pull, thereby reducing the space needed to mount sockets to an absolute minimum, up to 50 sockets per square decimeter.