Learn More
Abcam Streptavidin HRP (ready-to-use), 125ML
Supplier:  Abcam AB64269125ML
Encompass_Preferred
Streptavidin HRP (ready-to-use) (pH: 7.6) is a streptomyces avidinii protein and is suitable for immunohistochemistry (IHC). - Cited in over 45 publications - Contains PBS - Used with biotinylated secondary antibodies during IHC - Forms a strong, specific non-covalent complex with biotin (one per streptavidin subunit)
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0591436