Learn More
McMaster-Carr RTV159 Structural Silicone Sealant
Supplier:  McMaster-Carr 7589A23
Encompass
These structural sealants combine the mechanical strength of an adhesive with excellent sealing properties and high flexibility. They’re non-sagging, so they won't drip from vertical and overhead surfaces. Made from silicone, these sealants are RTV (room-temperature vulcanizing).
Catalog No. NC1773190