Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shelby™ 5284 Flex Structural Fire Gloves

Catalog No. 19100357 Shop All Shelby Specialty Gloves Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Jumbo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-100-357 Jumbo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-100-357 Supplier Shelby™ Supplier No. 5284J
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Durable koala tanned leather on the palm ensures ultra-soft, abrasion resistant protection. Insular textile construction on the back of the hand provides thermal and flame resistance without sacrificing flexibility.

Specifications

Certifications/Compliance NFPA 1971-2013
Cuff Material Nomex™
Color Yellow, Blue
Hand Specific
Material Aramid Fiber
Product Type Structural Firefighting Gloves
Size Jumbo
For Use With (Application) Structural Firefighting
Gender Unisex
Glove Cuff Color Black
Glove Cuff Style Knit
Palm Type Koala Tanned Leather
Series 5284
Length (English) Cuff 4 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.