Learn More
Molecular Dimensions STRUCTURE SCREEN I + II HT-96
SDP
Catalog No. NC0533063
Encompass
Protein Crystallography. Structure Screen 1 + 2 HT-96 (96 x 1.1 mL) is formulated for the crystallization of proteins, peptides, nucleic acids and water soluble small molecules.
Catalog No. NC0533063 Supplier Molecular Dimensions Supplier No. MD130
Protein Crystallography. Structure Screen 1 + 2 HT-96 (96 x 1.1 mL) is formulated for the crystallization of proteins, peptides, nucleic acids and water soluble small molecules.