Learn More
Grainger Hydra-Zorb Strut Mount Cushion Clamp, 1/2 in. OD, Carbon Steel, 1 ea.
Catalog No. 502785257
Encompass_Preferred
These strut mount cushion clamps are designed to provide fast, simple, and secure installation for tube and pipe runs. The assembly is engineered for use on uninsulated lines subject to damage from shock, movement, and vibration, effectively eliminating metal-to-metal contact between fluid conductors and clamps.
  • Features a carbon steel clamp with a thermoplastic elastomer cushion
  • Cushion material is rated for temperatures from -65°F to 275°F
  • Allows for addition or removal of fluid conductors without disturbing adjacent lines
  • Permits mixing of various size lines
  • Includes zinc-plated steel clamp, steel nylock nut, and steel weld stud
  • Complies with MSS SP-58 Type 61 and UL 2043 (25/50) standards
Catalog No. 50-278-5257 Supplier Grainger Supplier No. 193WK2
These strut mount cushion clamps are designed to provide fast, simple, and secure installation for tube and pipe runs. The assembly is engineered for use on uninsulated lines subject to damage from shock, movement, and vibration, effectively eliminating metal-to-metal contact between fluid conductors and clamps.
  • Features a carbon steel clamp with a thermoplastic elastomer cushion
  • Cushion material is rated for temperatures from -65°F to 275°F
  • Allows for addition or removal of fluid conductors without disturbing adjacent lines
  • Permits mixing of various size lines
  • Includes zinc-plated steel clamp, steel nylock nut, and steel weld stud
  • Complies with MSS SP-58 Type 61 and UL 2043 (25/50) standards