Learn More
Grainger ZSI Strut Mount Cushion Clamp, 3/4 in. OD Tube, Zinc Plated Steel, 25/Pk.
Catalog No. 502785308
Encompass_Preferred
This strut mount cushion clamp is designed for secure fluid conductor management and pipe or tubing support in industrial environments. It features a robust construction suitable for demanding applications, ensuring reliable performance and stability for 3/4 in. tubing.
  • Features a clamp manufactured from zinc plated steel
  • Includes a steel nylock nut and a steel weld stud
  • Operates within a temperature range of -65°F to 275°F
  • Engineered for use with standard support systems
Catalog No. 50-278-5308 Supplier Grainger Supplier No. 193WK4
This strut mount cushion clamp is designed for secure fluid conductor management and pipe or tubing support in industrial environments. It features a robust construction suitable for demanding applications, ensuring reliable performance and stability for 3/4 in. tubing.
  • Features a clamp manufactured from zinc plated steel
  • Includes a steel nylock nut and a steel weld stud
  • Operates within a temperature range of -65°F to 275°F
  • Engineered for use with standard support systems