Learn More
Grainger Nvent Caddy Fixed Strut Support, 6 in. x 4.8 in., Hot-Dip Galvanized, 1 ea.
Catalog No. 502786804
Encompass_Preferred
This fixed strut support provides a simple, adaptable solution for mounting electrical, mechanical, and HVAC services on commercial rooftops. It is engineered to distribute weight effectively across the base to ensure optimal load management while protecting roofing materials. The system features a durable, hot-dip galvanized strut profile and a UV-stabilized base.
  • Provides superior support for pipe, conduit, duct, cable tray, and equipment
  • Foam bottom offers a low-abrasion interface for enhanced roof membrane protection
  • Compatible with single-ply, bituminous, metal, and spray-foam roof surfaces
  • Ready to use out of the box to save installation time and labor
  • Constructed with UV-stabilized materials and hot-dip galvanized steel for corrosion resistance
  • Supports green building requirements with recycled thermoplastic base materials
Catalog No. 50-278-6804 Supplier Grainger Supplier No. 190A04
This fixed strut support provides a simple, adaptable solution for mounting electrical, mechanical, and HVAC services on commercial rooftops. It is engineered to distribute weight effectively across the base to ensure optimal load management while protecting roofing materials. The system features a durable, hot-dip galvanized strut profile and a UV-stabilized base.
  • Provides superior support for pipe, conduit, duct, cable tray, and equipment
  • Foam bottom offers a low-abrasion interface for enhanced roof membrane protection
  • Compatible with single-ply, bituminous, metal, and spray-foam roof surfaces
  • Ready to use out of the box to save installation time and labor
  • Constructed with UV-stabilized materials and hot-dip galvanized steel for corrosion resistance
  • Supports green building requirements with recycled thermoplastic base materials