Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strychnine Hydrochloride, Dihydrate, 98%, Spectrum™ Chemical
SDP

Catalog No. 18614119 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-119 25 g Amber Glass Bottle
18-614-117 100 g Amber Glass Bottle
18-614-118 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-614-119 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ST14525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6101-04-8

Specifications

Physical Form Crystals and/or Chunks
CAS Max % 1
Quantity 25 g
Packaging Amber Glass Bottle
Percent Purity 98%
Grade Ungraded
Chemical Name or Material Strychnine Hydrochloride, Dihydrate

CAUTION: For use by qualified personnel for research and development or other authorized use only. Not for food, drug or pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.