Learn More
Aviva Systems Biology Corp STS Antibody - C-terminal region (ARP41710_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41710_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against STS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human STS. Alternate names: ARSC, ARSC1, ASC, ES, SSDD, XLIApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-57963