Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STS-E412 | 1609980-39-3 | 99.7% | 318.76 | C15H15ClN4O2 | 1 ML
SDP

Catalog No. 5000210776
Encompass_Preferred

STS-E412 is a selective activator of the tissue-protective erythropoietin receptor complex (EPOR and CD131), supplied as a 10 mM solution in DMSO for preclinical research in neurodegenerative disease and organ protection.

  • Selective activator of tissue-protective EPO receptor
  • Supplied as a 10 mM solution in DMSO, 1 mL
  • High reported purity (99.7%)
  • Molecular weight 318.76; formula C15H15ClN4O2
  • Solubility: readily soluble in DMSO; in vivo formulation possible in 10% DMSO/90% corn oil
  • Recommended storage: powder -20°C (long term) or 4°C; in solvent -80°C (preferred) or -20°C

Catalog No. 50-002-10776 Supplier Medchemexpress LLC Supplier No. HYW06270010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STS-E412 is a selective activator of the tissue-protective erythropoietin receptor complex (EPOR and CD131), supplied as a 10 mM solution in DMSO for preclinical research in neurodegenerative disease and organ protection.

  • Selective activator of tissue-protective EPO receptor
  • Supplied as a 10 mM solution in DMSO, 1 mL
  • High reported purity (99.7%)
  • Molecular weight 318.76; formula C15H15ClN4O2
  • Solubility: readily soluble in DMSO; in vivo formulation possible in 10% DMSO/90% corn oil
  • Recommended storage: powder -20°C (long term) or 4°C; in solvent -80°C (preferred) or -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.