Learn More
Bioworld StAsis Stem Cell Qualified U.S. Origin Fetal Bovine Serum, 500 mL
Catalog No. 50254169
Encompass_Preferred
Stem cells are the least differentiated of all cell types. Many biomolecules common in sera can trigger unwanted differentiation. As such, these cells have uniquely pristine requirements for growth in culture.Our stem cell qualified fetal bovine serum has been specifically tested for its ability to sustain the undifferentiated cellular morphology of these cells. This quality product comes exclusively from US sources.SpecificationsTriple 0.1µm sterile-filteredMycoplasma-tested and virus-screenedShips on dry ice.
Catalog No. 50-254-169 Supplier Bioworld Supplier No. 306110702
Stem cells are the least differentiated of all cell types. Many biomolecules common in sera can trigger unwanted differentiation. As such, these cells have uniquely pristine requirements for growth in culture.Our stem cell qualified fetal bovine serum has been specifically tested for its ability to sustain the undifferentiated cellular morphology of these cells. This quality product comes exclusively from US sources.SpecificationsTriple 0.1µm sterile-filteredMycoplasma-tested and virus-screenedShips on dry ice.