Learn More
Enzo Life Sciences STT Negative Control for Gelatinase (5mg)
SDP
Supplier:  Enzo Life Sciences BMLPI1370005
Encompass_Preferred
STTHWGFTLS is ten times less potent than, thus a negative control for CTTHWGFTLC (BML-PI136), which is a specific inhibitor of the gelatinases MMP-2 and MMP-9. Can be used in in vitro studies, zymography and in situ zymography, in tissue culture, and in vivo. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water (10mg/ml) or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1474