Learn More
Cole-Parmer BH-200D-3-HT-120 Stuart Digital 3-Block Heater, 200C; 120 VAC
Catalog No. NC1936887 Shop All Cole-Parmer Products
Encompass
Dry block heaters are designed for precise heating of test tubes, microcentrifuge tubes, cuvettes, and microtiter plates. The large thermal mass of the aluminum blocks provides very accurate and stable temperature control. The number of blocks accepted indicates the number of single-width blocks accepted by each heater. A heater that accepts two single-width blocks will also accept one double-width block.
Catalog No. NC1936887 Supplier Cole-Parmer Supplier No. 3661022
Dry block heaters are designed for precise heating of test tubes, microcentrifuge tubes, cuvettes, and microtiter plates. The large thermal mass of the aluminum blocks provides very accurate and stable temperature control. The number of blocks accepted indicates the number of single-width blocks accepted by each heater. A heater that accepts two single-width blocks will also accept one double-width block.