Learn More
Cole-Parmer Stuart SCF3 Microcentrifuge, 500 to 15,000 rpm; 100 to 240 VAC, 50/60 Hz
Catalog No. 501960448 Shop All Cole-Parmer Products
Encompass_Preferred
This microcentrifuge is small and quiet enough for personal use, but offers advanced features typical of larger benchtop centrifuges. In addition to the user-friendly digital interface, the centrifuge comes with software and a USB cable allowing for remote operation and data acquisition. For safety, the lid remains locked while the rotor is in motion and automatically releases when the rotor comes to a halt. In the event of a power failure, an emergency lid release latch is installed in the base. Program speed or RCF and time, which is saved in onboard memory. Imbalance detection system identifies an asymmetrical load and prevents operation.
Catalog No. 50-196-0448 Supplier Cole-Parmer Supplier No. 8523000
This microcentrifuge is small and quiet enough for personal use, but offers advanced features typical of larger benchtop centrifuges. In addition to the user-friendly digital interface, the centrifuge comes with software and a USB cable allowing for remote operation and data acquisition. For safety, the lid remains locked while the rotor is in motion and automatically releases when the rotor comes to a halt. In the event of a power failure, an emergency lid release latch is installed in the base. Program speed or RCF and time, which is saved in onboard memory. Imbalance detection system identifies an asymmetrical load and prevents operation.