Learn More
Electron Microscopy Sciences Tripod Polisher Stub Assembly
SDP
Catalog No. 50980881
Encompass
The SEM Stub is used for mounting samples for SEM and can be transferred directly to the SEM or to the special holder for use in mil. - For use with Tripod Polisher Model 590.
Catalog No. 50-980-881 Supplier Electron Microscopy Sciences Supplier No. 5011665
The SEM Stub is used for mounting samples for SEM and can be transferred directly to the SEM or to the special holder for use in mil. - For use with Tripod Polisher Model 590.