Learn More
Aviva Systems Biology Corp STUB1 Antibody - N-terminal region (ARP43058_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43058_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STUB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STUB1. Alternate names: CHIP, HSPABP2, NY-CO-7, SDCCAG7, UBOX1ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58411